Learn More
Dräger™ CMS Chips: Perchloroethylene
Product Code. 10321022
Change view
Click to view available options
Measurement Range:
5 to 500
Unit Size:
Each
Product Code. Measurement Range Unit Size
10321022 5 to 500 Each
1 options
This item is not returnable. View return policy
Product Code. 10321022 Supplier Dräger™ Supplier No. 6406040
  • ONLINE EXCLUSIVE PRICE
    244.95€
    202.64€ / paquete de 1
    Ahorro 42.31€
    17% Descuento
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Perchloroethylene CMS Chip
Humidity Range 5 to 30mg/L
Measurement Range 5 to 500
Response Time 40 to 330 sec.
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15° to 40°C