Learn More
Dräger™ CMS Chips: Ammonia
Product Code. 12394599
Change view
Click to view available options
Measurement Range:
0.2 to 5
10 to 150
2 to 50
Response Time:
100 to 600 sec.
15 to 140 sec.
15 to 50 sec.
Humidity Range:
1 to 30mg/L
1 to 40mg/L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Response Time Humidity Range Unit Size
12394599 0.2 to 5 100 to 600 sec. 1 to 30mg/L Each
10426301 2 to 50 15 to 140 sec. 1 to 40mg/L Each
10436301 10 to 150 15 to 50 sec. 1 to 40mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12394599 Supplier Dräger™ Supplier No. 6406550
  • ONLINE EXCLUSIVE PRICE
    320.57€
    265.55€ / paquete de 1
    Ahorro 55.02€
    17% Descuento
Fecha estimada de envoi: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Ammonia CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 0.2 to 5
Response Time 100 to 600 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C