Learn More
Dräger™ CMS Chips: Carbon Monoxide
Product Code. 10416301
Change view
Click to view available options
Measurement Range:
5 to 150
Unit Size:
Each
Product Code. Measurement Range Unit Size
10416301 5 to 150 Each
1 options
This item is not returnable. View return policy
Product Code. 10416301 Supplier Dräger™ Supplier No. 6406080
  • ONLINE EXCLUSIVE PRICE
    219.39€
    181.47€ / paquete de 1
    Ahorro 37.92€
    17% Descuento
Fecha estimada de envoi: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Carbon Monoxide CMS Chip
Humidity Range 1 to 50mg/L
Measurement Range 5 to 150
Response Time 80 to 300 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C