Learn More
Dräger™ CMS Chips: Ethanol
Product Code. 10287467
Change view
Click to view available options
Measurement Range:
100 to 2500
Unit Size:
Each
Product Code. Measurement Range Unit Size
10287467 100 to 2500 Each
1 options
This item is not returnable. View return policy
Product Code. 10287467 Supplier Dräger™ Supplier No. 6406370
  • ONLINE EXCLUSIVE PRICE
    259.86€
    215.45€ / paquete de 1
    Ahorro 44.41€
    17% Descuento
 Restricted Item
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Ethanol CMS Chip
Humidity Range 5 to 25mg/L
Measurement Range 100 to 2500
Response Time 60 to 340 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C
compliance-icons
Product Identifier
  • GAS SAMPLING CASSETTE DRAEGER CMS CHIP ETHANOL 10
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid