Learn More
Dräger™ CMS Chips: Acetic Acid
Product Code. 12865023
Change view
Click to view available options
Measurement Range:
2 to 50
Unit Size:
Each
Product Code. Measurement Range Unit Size
12865023 2 to 50 Each
1 options
This item is not returnable. View return policy
Product Code. 12865023 Supplier Dräger™ Supplier No. 6406330
  • ONLINE EXCLUSIVE PRICE
    233.24€
    193.41€ / paquete de 1
    Ahorro 39.83€
    17% Descuento
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Acetic Acid CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 2 to 50
Response Time 40 to 300 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C