missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DPYD (Human) Recombinant Protein
Human DPYD full-length ORF ( AAH08379, 1 a.a. - 173 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001806-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAPVLSKDSADIESILALNPRTQTHATLCSTSAKKLDKKHWKRNPDKNCFNCEKLENNFDDIKHTTLGERGALREAMRCLKCADAPCQKSCPTNLDIKSFITSIANKNYYGAAKMIFSDNPLGLTCGMVCPTSDLCVGGCNLYATEEGPINIGGLQQFATETLILAFSLMNHL
Especificaciones
AAH08379 | |
dihydropyrimidine dehydrogenase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DPYD | |
GST |
1806 | |
Wheat germ expression system | |
25 μg | |
DHP, DHPDHASE, DPD, MGC132008, MGC70799 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |