missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPH2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12367-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody works well in WB, IP, IHC and IF application.
This gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamide mutant strain, and thus probably functions in diphthamide biosynthesis. Diphthamide is a post-translationally modified histidine residue present in elongation factor 2 (EF2) that is the target of diphtheria toxin ADP-ribosylation. Two transcript variants encoding different isoforms have been found for this gene.Especificaciones
DPH2 | |
Polyclonal | |
Unconjugated | |
DPH2 | |
DPH2, DPH2 homolog, DPH2 homolog (S. cerevisiae), DPH2 like 2, DPH2L2, HsDph2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1802, 67728 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BQC3, Q9CR25 | |
DPH2 | |
DPH2 Fusion Protein Ag3042 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |