missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DPH1 (Human) Recombinant Protein
Human DPH1 full-length ORF ( NP_001374, 1 a.a. - 363 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001801-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MPEGLLLFACTIVDILERFTEAEVMVMGDVTYGACCVDDFTARALGADFLVHYGHSCLIPMDTSAQDFRVLYVFVDIRIDTTHLLDSLRLTFPPATALALVSTIQFVSTLQAAAQELKAEYRVSVPQCKPLSPGEILGCTSPRLSKEVEAVVYLGDGRFHLESVMIANPNVPAYRYDPYSKVLSREHYDHQRMQAARQEAIATARSAKSWGLILGTLGRQGSPKILEHLESRLRALGLSFVRLLLSEIFPSKLSLLPEVDVWVQVACPRLSIDWGTAFPKPLLTPYEAAVALRDISWQQPYPMDFYAGSSLGPWTVNHGQDRRPHAPGRPARGKVQEGSARPPSAVACEDCSCRDEKVAPLAP
Especificaciones
NP_001374 | |
DPH1 homolog (S. cerevisiae) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DPH1 | |
GST |
1801 | |
Wheat germ expression system | |
25 μg | |
DPH2L, DPH2L1, FLJ33211, OVCA1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |