Learn More
Dopamine 4-Beta-D-Glucuronide, TRC
Product Code. 30138285
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138285 10 mg 10mg
30011413 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138285 Supplier Toronto Research Chemicals Supplier No. TRCD53379510MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38632-24-5
Molecular Formula C14H19NO8
Molecular Weight (g/mol) 329.3
Synonym 4-(2-Aminoethyl)-2-hydroxyphenyl β-D-Glucopyranosiduronic Acid, Dopamine Glucuronide, Dopamine-4-O-glucuronide;
SMILES OC1=C(O[C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O2)C=CC(CCN)=C1
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C14H19NO8/c15-4-3-6-1-2-8(7(16)5-6)22-14-11(19)9(17)10(18)12(23-14)13(20)21/h1-2,5,9-12,14,16-19H,3-4,15H2,(H,20,21)/t9-,10-,11+,12-,14+/m0/s1
Formula Weight 329.1111
Chemical Name or Material Dopamine 4-beta-D-Glucuronide