Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody, Texas Red, Novus Biologicals™
Donkey Polyclonal Antibody
Marca:  Bio-Techne NB7600
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215414
  • 227.86€ / 1 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Especificaciones
Especificaciones
IgG (H+L)
Polyclonal
Western Blot 1:1000 - 1:10000, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Donkey
Multi-step
RUO
Secondary
Mouse
Purified
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Texas Red
IGHG
Mouse IgG whole molecule
1 mg
Adaptive Immunity, Immunology
Mouse IgG (H and L)
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only