Learn More
Thermo Scientific™ Dolly for 300L Plastic Drum for HyPerforma™ Single-Use Mixer DS 300 (stainless steel)
Product Code. 17958211 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
17958211 1 Ea. Each
1 options
This item is not returnable. View return policy
Product Code. 17958211 Supplier Thermo Scientific™ Supplier No. SH30958.03
  • ONLINE EXCLUSIVE PRICE
    597.81€
    526.34€ / paquete de 1
    Ahorro 71.47€
    11% Descuento
 Restricted Item
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Dolly (single pack) is designed for use with 300L Plastic Drums and the Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.) DS 300. Dollies for the drums are available in two sizes: one for 50 to 200L drums and another for 300L drums. These drums should be used with the corresponding optional drum positioner that is available with the docking station.
The Dolly (single pack) is designed for use with 300L Plastic Drums and the Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.) DS 300. Dollies for the drums are available in two sizes: one for 50 to 200L drums and another for 300L drums. These drums should be used with the corresponding optional drum positioner that is available with the docking station.
Required accessories:
  • HyPerforma Single-Use Mixer DS 300 docking station
  • HyPerforma Single-Use Mixer DS 300 bioprocess container bags for closed-top mixing or liners for open-top mixing
Recommended for:
  • Powder-liquid or liquid-liquid mixing applications in single-use
  • High-throughput mixing and storage
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) DS 300 Single-Use Mixer
Capacity (Metric) 300 L
Product Line HyPerforma
Material Polyethylene
Quantity 1 Ea.