Learn More
Thermo Scientific™ Dog (Beagle) Cytosol
Liver cytosolic fractions prepared from a pool of male Beagle Dogs (total protein concentration: 20mg/mL).
Marca: Thermo Scientific™ DGCYPL
Detalles adicionales : Peso : 0.99000kg
Descripción
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
Especificaciones
20mg/mL | |
Research Grade | |
Pooled | |
RUO - research use only | |
Canine |
Target-Based ADME/Tox Assays, ADME Assay, Enzymes and Proteins for ADME, Hepatocytes and Hepatic Models, Industrial and Applied Science, Pharma and Biopharma | |
1mL | |
Store between -60°C and -85°C. | |
Tube(s) | |
Frozen |