Learn More
DNAJC15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16063-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
DNAJC15 is a negative regulator of the mitochondrial respiratory chain. It prevents mitochondrial hyperpolarization state and restricts mitochondrial generation of ATP. It acts as an import component of the TIM23 translocase complex. It stimulates the ATPase activity of HSPA9.Especificaciones
DNAJC15 | |
Polyclonal | |
Unconjugated | |
DNAJC15 | |
DNAJC15, DNAJD1, HSD18, MCJ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290370, 29103, 66148 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q78YY6, Q9Y5T4 | |
Dnajc15 | |
DNAJC15 Fusion Protein Ag8911 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |