missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLEC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20027-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene contains 37 exons, spans approximately 59-kb, and is located in the 3p22-p21.3 chromosomal segment that is commonly deleted in various carcinomas. Several alternatively spliced transcripts have been observed that contain disrupted coding regions and likely encode nonfunctional proteins. Aberrant transcription of this gene may be involved in carcinogenesis of the lung, esophagus, and kidney.Especificaciones
DLEC1 | |
Polyclonal | |
Unconjugated | |
DLEC1 | |
DLC 1, DLC1, DLEC1, F56 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
320256, 9940 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BLA1, Q9Y238 | |
Dlec1 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |