Learn More
DL-2,4-Diaminobutyric Acid Dihydrochloride, TRC
Product Code. 30386928
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386928 10 mg 10mg
30381172 100 mg -
30384744 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386928 Supplier Toronto Research Chemicals Supplier No. TRCD45355310MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65427-54-5
Molecular Formula C4H10N2O2 . 2HCl
Molecular Weight (g/mol) 191.05
Synonym Butanoic acid, 2,4-diamino-, dihydrochloride (9CI), Butanoic acid, 2,4-diamino-, dihydrochloride, (±)-, 2,4-Diaminobutanoic acid dihydrochloride, 2,4-Diaminobutyric acid dihydrochloride, NSC 41117, dl-2,4-Diaminobutyric acid dihydrochloride
SMILES Cl.Cl.NCCC(N)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C4H10N2O2.2ClH/c5-2-1-3(6)4(7)8;;/h3H,1-2,5-6H2,(H,7,8);2*1H
Formula Weight 190.0276
Chemical Name or Material DL-2,4-Diaminobutyric Acid Dihydrochloride