Learn More
Lenz Laborglasintrumente Distribution Tubes With Frit
Product Code. 10681892
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
For Use With (Application):
Specific heavy solvents
Specific light solvents
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) For Use With (Application) Unit Size
10681892 100 mL Specific heavy solvents Each
10204151 250 mL Specific heavy solvents Each
10264001 250 mL Specific light solvents Each
10306121 100 mL Specific light solvents Each
10336071 500 mL Specific light solvents Each
10336311 500 mL Specific heavy solvents Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10681892 Supplier Lenz Laborglasintrumente Supplier No. 05433202
  • ONLINE EXCLUSIVE PRICE
    56.00€
    37.55€ / paquete de 1
    Ahorro 18.45€
    32% Descuento
Fecha estimada de envoi: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Distribution tubes with frit (porosity: 0)
  • For specific light solvents
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 100 mL
For Use With (Application) Specific heavy solvents