Learn More
3M™ Zeta Plus SP Series Scale-Up Filter Capsule
Marca: 3M™ 7000042853
Detalles adicionales : Peso : 0.10000kg
Especificaciones
10SP02A | |
170.3 cm2 | |
1Case | |
Scale-Up Filter Capsule |
Cellulose | |
Adsorptive Depth Filtration | |
1Case | |
10.6 cm |