missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Dispensette™ S Analog-adjustable Bottletop Dispensers, Exchange Program

Features a 'floating piston' design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.

Marca:  Brand™ 4600501

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933440

  • 224.15€ / cada uno


Fecha estimada de envío
08-07-2020
Añadir a la cesta

Descripción

Descripción

  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Safe volume setting with internal scalloped track (analog-adjustable instruments)
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking
  • Exchange promotion
Especificaciones

Especificaciones

A 121 °C
Rojo
Con válvula de recirculación
De 0.1 a 1 ml
Aplicaciones: Dispensación de reactivos agresivos, incluidos ácidos y bases concentrados, como H3PO4, H2SO4 (excepto, por ejemplo, HCl, HNO3 y HF), soluciones salinas, además de numerosos disolventes orgánicos
0.6% (6μL)
ISO 9001
Dispensador
Analógico
0.02 mL
Videos
Documentos