Learn More
Diphenhydramine N-Beta-D-Glucuronide, TRC
Product Code. 30012567
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012567 25 mg 25mg
30004376 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012567 Supplier Toronto Research Chemicals Supplier No. TRCD48691025MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 137908-78-2
Molecular Formula C23 H29 N O7
Molecular Weight (g/mol) 431.48
Synonym Ethanaminium, 2-(diphenylmethoxy)-N-β-D-glucopyranuronosyl-N,N-dimethyl-, inner salt, Diphenhydramine N-glucuronide, Diphenhydramine N-β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-6-[2-benzhydryloxyethyl(dimethyl)azaniumyl]-3,4,5-trihydroxyoxane-2-carboxylate
SMILES C[N+](C)(CCOC(c1ccccc1)c2ccccc2)[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-]
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C23H29NO7/c1-24(2,22-19(27)17(25)18(26)21(31-22)23(28)29)13-14-30-20(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,17-22,25-27H,13-14H2,1-2H3/t17-,18-,19+,21-,22+/m0/s1
Formula Weight 431.1944
Chemical Name or Material Diphenhydramine N-beta-D-Glucuronide