Learn More
Lenz Laborglasintrumente Dimroth Condensers, with Glass Olives
Product Code. 10139600
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 24/29
Length (Metric) Jacket:
160 mm
250 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
10139600 NS 14/23 250 mm Each
10047770 NS 14/23 160 mm Each
10101621 NS 24/29 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10139600 Supplier Lenz Laborglasintrumente Supplier No. 05240301
  • ONLINE EXCLUSIVE PRICE
    141.00€
    119.10€ / paquete de 1
    Ahorro 21.90€
    15% Descuento
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Dimroth condensers acc. to DIN 12591
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Includes Glass olives
Length (Metric) Jacket 250 mm
Quantity 1 Ea.
Standard Taper Joint No NS 14/23
Material Duran Glass