missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimetilglioxima, ACS, Thermo Scientific Chemicals
Click to view available options
Cantidad:
50 g
250 g
Descripción
Dimethylglyoxime, is used as a chelating agent which forms complexes with metals such as copper, nickel, and palladium. Dimethylglyoxime reacts with a nickel salt in a green solution to generate a red colored insoluble tetradentate coordinate complex of nickel. It is used as a reagent for the colorimetric determination of urea and ureido compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Dimethylglyoxime |
Punto de fusión | ∼240°C (decomposition) |
Color | Blanco |
Cantidad | 250 g |
Beilstein | 506731 |
Índice Merck | 14,3246 |
Información de solubilidad | Soluble in alcohol, acetone, ether |
Formula Weight (peso de la fórmula) | 116.12 |
Grado | ACS |
Forma física | Polvo |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'