Learn More
Dicloroacetato de diisopropilamina, 95 %, Thermo Scientific Chemicals
Catalog Number 15447868
Click to view available options
Cantidad:
250 mg
1 g
5 g
660-27-5
C8H17Cl2NO2
230.13
MFCD00868288
ILKBHIBYKSHTKQ-UHFFFAOYSA-N
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
12617
ácido 2,2-dicloroacético; N-propan-2-ilpropan-2-amina
OC(=O)C(Cl)Cl.CC(C)NC(C)C
660-27-5
C8H17Cl2NO2
230.13
MFCD00868288
ILKBHIBYKSHTKQ-UHFFFAOYSA-N
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
12617
ácido 2,2-dicloroacético; N-propan-2-ilpropan-2-amina
OC(=O)C(Cl)Cl.CC(C)NC(C)C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Diisopropylamine dichloroacetate
Color Blanco
Fórmula lineal [(CH3)2CH]2NH·Cl2CHCO2H
Cantidad 5 g
Formula Weight (peso de la fórmula) 230.13
Porcentaje de pureza 95%
Forma física Polvo cristalino