Learn More
Diethylene glycol monoethyl ether acetate, 99%
Beskrivelse
Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with bases and oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Tekniske data
Tekniske data
| Melting Point | -25°C |
| Density | 1.012 |
| Boiling Point | 218°C to 219°C |
| Flash Point | 95°C (203°F) |
| Refractive Index | 1.421 |
| Quantity | 2500 mL |
| Beilstein | 1764643 |
| Merck Index | 14,1800 |
| Solubility Information | Soluble in water. |
| Formula Weight | 176.21 |
| Vis mere |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.