missing translation for 'onlineSavingsMsg'
Learn More

Diethyl oxalacetate sodium salt, 95%

Product Code. 11487024
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487024 100 g 100g
11497024 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487024 Supplier Thermo Scientific Alfa Aesar Supplier No. B21486.22
 Restricted Item
En stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40876-98-0 | C8H11NaO5 | 210.161 g/mol

Diethyl oxalacetate sodium salt is used in the preparation of oxaloacetic acid. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl oxalacetate sodium salt is used in the preparation of oxaloacetic acid. It is also used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water (1.2 g/L) (25°C).

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40876-98-0
Molecular Formula C8H11NaO5
Molecular Weight (g/mol) 210.161
MDL Number MFCD00035571
InChI Key UJZUICGIJODKOS-UHFFFAOYSA-M
Synonym diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
PubChem CID 44134875
IUPAC Name sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
SMILES CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]

Specifications

Melting Point ∼185°C
Quantity 100 g
Beilstein 4279609
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water (1.2g/L) (25°C).
Formula Weight 210.16
Percent Purity 95%
Chemical Name or Material Diethyl oxalacetate sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.