missing translation for 'onlineSavingsMsg'
Learn More

Dietilo metilfosfonita, Thermo Scientific Chemicals

Catalog Number p-7049856
Click to view available options
Cantidad:
0.5 g
2 g
10 g
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: es posible que la compra de este producto esté sujeta a restricciones y necesitemos solicitarle documentación adicional.
15715-41-0
C5H13O2P
136.13
MFCD00009089
NSSMTQDEWVTEKN-UHFFFAOYSA-N
diethyl methylphosphonite, methyldiethoxyphosphine, phosphonous acid, methyl-, diethyl ester, diethoxy methyl phosphane, o,o-diethyl methylphosphonite, diethyl methanephosphonite, phosphonous acid, p-methyl-, diethyl ester, acmc-20akel, diethoxymethylphosphine, diethylmethylphosphonite
85063
dietoxi(metil)fosfano
CCOP(C)OCC
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: es posible que la compra de este producto esté sujeta a restricciones y necesitemos solicitarle documentación adicional.
15715-41-0
C5H13O2P
136.13
MFCD00009089
NSSMTQDEWVTEKN-UHFFFAOYSA-N
diethyl methylphosphonite, methyldiethoxyphosphine, phosphonous acid, methyl-, diethyl ester, diethoxy methyl phosphane, o,o-diethyl methylphosphonite, diethyl methanephosphonite, phosphonous acid, p-methyl-, diethyl ester, acmc-20akel, diethoxymethylphosphine, diethylmethylphosphonite
85063
dietoxi(metil)fosfano
CCOP(C)OCC

Dimethyl methylphosphonate is a phosphonite reagent used in the generation of phosphinic acid derivative via Arbuzov reaction and as a cyclization reagentin the preparation of 2-substituted penems. It is a chiral intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Diethyl methylphosphonite
Olor Strong
Índice de refracción 1.4168
Cantidad 0.5 g
Número UN UN3279
Sensibilidad Air and moisture sensitive
Información de solubilidad It slowly hydrolyses in water.
Formula Weight (peso de la fórmula) 136.13
Forma física Líquido

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'