Learn More
Alfa Aesar™ Diethyl ethylphosphonate, 98%
Marca: Alfa Aesar™ A11046.18
Detalles adicionales : CAS : 78-38-6 Peso : 0.05000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Cantidad. | 50g |
---|
Descripción
ApplicationsDiethyl ethylphosphonate is used for heavy-metal extraction and solvent separation. It is also used as a gasoline additive, antifoam agent, plasticizer and textile conditioner. Further, it serves as an antistatic agent, flame retardant and as a general reagent in phosphinoalkene and phosphinoalkyne synthesis.
Solubility
Miscible with alcohol, ether and organic solvents. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Identificadores de productos químicos
78-38-6 | |
166.157 | |
AATNZNJRDOVKDD-UHFFFAOYSA-N | |
6534 | |
CCOP(=O)(CC)OCC |
C6H15O3P | |
MFCD00039888 | |
diethyl ethylphosphonate, phosphonic acid, ethyl-, diethyl ester, diethyl ethanephosphonate, ethanephosphonic acid, diethyl ester, unii-y52q01p44t, ccris 6230, diethylethanephosphonate, ethylphosphonic acid diethyl ester, phosphonic acid, p-ethyl-, diethyl ester, diethylethylphosphonate | |
1-[ethoxy(ethyl)phosphoryl]oxyethane |
Especificación
Diethyl ethylphosphonate | |
82°C to 83°C (11mmHg) | |
78-38-6 | |
MFCD00039888 | |
1756861 | |
AATNZNJRDOVKDD-UHFFFAOYSA-N | |
1-[ethoxy(ethyl)phosphoryl]oxyethane | |
6534 | |
98% |
105°C (221°F) | |
50g | |
C6H15O3P | |
UN3082 | |
diethyl ethylphosphonate, phosphonic acid, ethyl-, diethyl ester, diethyl ethanephosphonate, ethanephosphonic acid, diethyl ester, unii-y52q01p44t, ccris 6230, diethylethanephosphonate, ethylphosphonic acid diethyl ester, phosphonic acid, p-ethyl-, diethyl ester, diethylethylphosphonate | |
CCOP(=O)(CC)OCC | |
166.157 | |
166.16 |