Learn More
Alfa Aesar™ Diethyl allylphosphonate, 96%
Marca:  Alfa Aesar™ 010365.06
Detalles adicionales : CAS : 1067-87-4 Peso : 0.00500kg
Cantidad. 5g
 Ver más versiones de este producto
Código de producto. 11375818
  • 121.00€ / 5 g
Fecha de envío estimada
del proveedor 20-08-2019
Añadir a la cesta
Especificaciones
1067-87-4
178.168
YPJHXRAHMUKXAE-UHFFFAOYSA-N
573014
CCOP(=O)(CC=C)OCC
C7H15O3P
MFCD00015134
diethyl allylphosphonate, diethyl prop-2-en-1-ylphosphonate, diethyl-allylphosphonate, acmc-1bprn, phosphonic acid, 2-propenyl-, diethyl ester, allylphosphonic acid diethyl, diethyl allylphosphonate, 3-diethoxyphosphoryl-prop-1-ene, allylphosphonic acid diethyl ester, allyl phosphonic acid diethyl ester
3-diethoxyphosphorylprop-1-ene
Especificaciones
Diethyl allylphosphonate
>61° (141°F)
63°C (2.5mmHg)
1067-87-4
MFCD00015134
YPJHXRAHMUKXAE-UHFFFAOYSA-N
3-diethoxyphosphorylprop-1-ene
573014
Liquid
1.0356g/mL
1.4350
5g
C7H15O3P
diethyl allylphosphonate, diethyl prop-2-en-1-ylphosphonate, diethyl-allylphosphonate, acmc-1bprn, phosphonic acid, 2-propenyl-, diethyl ester, allylphosphonic acid diethyl, diethyl allylphosphonate, 3-diethoxyphosphoryl-prop-1-ene, allylphosphonic acid diethyl ester, allyl phosphonic acid diethyl ester
CCOP(=O)(CC=C)OCC
178.168
178.17
96%
Videos
SDS Documents