Learn More
Dietoximetano, 99 %, estab. con 100 ppm de BHT, Thermo Scientific Chemicals
Catalog Number p-7026863
Click to view available options
Cantidad:
25 mL
100 mL
500 mL
462-95-3
C5H12O2
104.15
MFCD00009246
KLKFAASOGCDTDT-UHFFFAOYSA-N
diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
10024
ethoxymethoxyethane
CCOCOCC
462-95-3
C5H12O2
104.15
MFCD00009246
KLKFAASOGCDTDT-UHFFFAOYSA-N
diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
10024
ethoxymethoxyethane
CCOCOCC
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Diethoxymethane
Nombre de nota stabilized with 100ppm BHT
Punto de fusión -66°C
Punto de ebullición 87°C to 88°C
Temperatura de inflamación −5°C (23°F)
Intervalo de porcentaje de ensayo 0.99
Cantidad 25 mL
Número UN UN2373
Beilstein 1697253
Formula Weight (peso de la fórmula) 104.15
Porcentaje de pureza 99%
Forma física Liquid