Learn More
Dibromo-tyrosine Antibody (9F12), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NBP2-59351AF700
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214555
  • 677.09€ / 100 microlitros
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
Dibromo-tyrosine Monoclonal specifically detects Dibromo-tyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
Dibromo-tyrosine
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for 3,5-Dibromo-tyrosine. Does not cross-react with Nitrotyrosine.
Store at 4C in the dark.
IgG1
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
9F12
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Mouse
Protein G purified
Primary
Non-species specific
Purified