Learn More
Ácido (-)-dibenzoil- D-tartárico monohidrato, 99 %, Thermo Scientific Chemicals
Catalog Number p-7023195
Click to view available options
Cantidad:
25 g
100 g
80822-15-7
C18H16O9
376.32
MFCD00150722
DXDIHODZARUBLA-DZWQFRRJNA-N
2s,3s-2,3-bis benzoyloxy succinic acid hydrate, +-dibenzoyl-d-tartaric acid monohydrate, d-dbta, dibenzoyl-d-+-tartaric acid monohydrate, o,o'-dibenzoyl d-tartaric acid monohydrate, +-2,3-dibenzoyl-d-tartaric acid monohydrate, +-dibenzoyl-d-tartaric acid, 2s,3s-+-dibenzoyl-d-tartaric acid monohydrate, 2s,3s-+-o,o'-dibenzoyltartaric acid monohydrate, butanedioic acid, 2,3-bis benzoyloxy-, monohydrate, 2s,3s
11667949
O.OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O
80822-15-7
C18H16O9
376.32
MFCD00150722
DXDIHODZARUBLA-DZWQFRRJNA-N
2s,3s-2,3-bis benzoyloxy succinic acid hydrate, +-dibenzoyl-d-tartaric acid monohydrate, d-dbta, dibenzoyl-d-+-tartaric acid monohydrate, o,o'-dibenzoyl d-tartaric acid monohydrate, +-2,3-dibenzoyl-d-tartaric acid monohydrate, +-dibenzoyl-d-tartaric acid, 2s,3s-+-dibenzoyl-d-tartaric acid monohydrate, 2s,3s-+-o,o'-dibenzoyltartaric acid monohydrate, butanedioic acid, 2,3-bis benzoyloxy-, monohydrate, 2s,3s
11667949
O.OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O
(+)-Dibenzoyl-D-tartaric acid monohydrate is used as a chiral separation and racemization agent for amines. It is also used as a pharmaceutical raw material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+)-Dibenzoyl-D-tartaric acid monohydrate is used as a chiral separation and racemization agent for amines. También se utiliza como materia prima farmacéutica.
Solubilidad
Soluble en metanol. Ligeramente soluble en agua.
Notas
incompatible con agentes oxidantes fuertes, bases y ácidos fuertes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (+)-Dibenzoyl-D-tartaric acid monohydrate
Punto de fusión De 87 °C a 91 °C
Beilstein 6121616
Información de solubilidad Soluble in methanol. Slightly soluble in water.
Nombre IUPAC (2S,3S)-2,3-bis(benzoyloxy)butanedioic acid hydrate
Formula Weight (peso de la fórmula) 376.33 (358.31 Anhydrous)
Porcentaje de pureza 99%
Cantidad 25 g
Rotación óptica +109° (c=2 en etanol)