Learn More
Diazolidinyl Urea (Technical Grade), TRC
Product Code. 30351276
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351276 50 g 50g
30360924 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30351276 Supplier Toronto Research Chemicals Supplier No. TRCD41732350G
 Restricted Item
Fecha estimada de envoi: 15-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78491-02-8
Molecular Formula C8H14N4O7
Molecular Weight (g/mol) 278.22
Synonym N-[1,3-Bis(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-N,N'-bis(hydroxymethyl)urea, Diazolidinylurea, Germaben II-E, Germall II, N-Hydroxymethyl-N-[1,3-di(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-N'-hydroxymethylurea
IUPAC Name 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
SMILES OCNC(=O)N(CO)C1N(CO)C(=O)N(CO)C1=O
Specifications
Quantity 50 g
InChI Formula InChI=1S/C8H14N4O7/c13-1-9-7(18)10(2-14)5-6(17)12(4-16)8(19)11(5)3-15/h5,13-16H,1-4H2,(H,9,18)
Formula Weight 278.0862
Chemical Name or Material Diazolidinyl Urea (Technical Grade)