missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ DexPure Vinyl Gloves

Catalog Number p-8010166
Click to view available options
Tamaño:
Small
Medium
Large
X-Large
XL
En polvo:
Con talco
Powdered
Color:
Azul
Translucent
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

Convenient and disposable, these translucent vinyl gloves can be used for a variety of tasks from the laboratory to the kitchen. Honeywell Safety Products™ DexPure Vinyl Gloves also have a smooth finish that provides great dexterity.

  • Translucent vinyl glove.
  • Smooth finish for great dexterity.
  • Certified for food contact and treated to prevent bacteria.
  • Suitable for the following:
    • Product inspection.
    • Precision assembly.
    • Laboratory work.
    • Handling of paints, inks and dyes.
  • Single use.
  • AQL:1.5.
TRUSTED_SUSTAINABILITY
Certificaciones/conformidad CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 374-2003 Chemical hazards, EN 455-1&2 Medical Standards, European directive n°1934/2004 : Food contact, EC Category PPE 3, EC Certificate Number 0075/797/162/11/08/0465, EC Attestation Number 0075/797/162/11/08/0465
Clasificación Dexterity: 5
Descripción AQL:1.5
Coating Material Vinyl
Color Translucent
Desechables Single-use
Material Vinyl
En polvo Powdered
Tipo de producto Gloves
Tamaño Large
Grosor (métrico) 0.1 mm
Para utilizar con (aplicación) Automotive, Aviation, Catering, Chemical, Food, Industrial Cleaning, Laboratory, Maintenance, Medical, Pharmaceutical, Petrochemical, Printing and Water Treatment industries.
Cantidad 100/PK
Longitud (métrico) 25 cm
Mostrar más Mostrar menos
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'