missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ DexPure Vinyl Gloves
Click to view available options
Tamaño:
Small
Medium
Large
X-Large
XL
En polvo:
Con talco
Powdered
Color:
Azul
Translucent
Descripción
- Translucent vinyl glove.
- Smooth finish for great dexterity.
- Certified for food contact and treated to prevent bacteria.
- Suitable for the following:
- Product inspection.
- Precision assembly.
- Laboratory work.
- Handling of paints, inks and dyes.
- Single use.
- AQL:1.5.
Especificaciones
Certificaciones/conformidad | CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 374-2003 Chemical hazards, EN 455-1&2 Medical Standards, European directive n°1934/2004 : Food contact, EC Category PPE 3, EC Certificate Number 0075/797/162/11/08/0465, EC Attestation Number 0075/797/162/11/08/0465 |
Clasificación | Dexterity: 5 |
Descripción | AQL:1.5 |
Coating Material | Vinyl |
Color | Translucent |
Desechables | Single-use |
Material | Vinyl |
En polvo | Powdered |
Tipo de producto | Gloves |
Tamaño | Medium |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'