Learn More
JULABO™ Descaling Agent
Product Code. 17198528
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
17198528 1 L Each
1 options
This item is not returnable. View return policy
Product Code. 17198528 Supplier JULABO™ Supplier No. 9940200
  • ONLINE EXCLUSIVE PRICE
    47.75€
    43.00€ / paquete de 1
    Ahorro 4.75€
    9% Descuento
 Restricted Item
Fecha estimada de envoi: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
JULABO Descaling Agent is for gentle and sanitary removal of lime residue.
  • Descaling Agent for gentle and sanitary removal of mineral deposits including lime residue
Specifications
Volume (Metric) 1 L
For Use With (Application) Removal of Lime Residue
compliance-icons
Product Identifier
  • descaling Agent (1 Liter)
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sulphamidic acid, Talgtrimethylammoniumchloride, propan-2-ol