missing translation for 'onlineSavingsMsg'
Learn More

DEFA6 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17923-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817934

  • 109.00€ / 20 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes appear to be clustered on chromosome 8.
Especificaciones

Especificaciones

DEFA6
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q01524
DEF6, DEFA6, Defensin 6, Defensin, alpha 6, defensin, alpha 6, Paneth cell, HD 6
Rabbit
IgG
20 μL
-20°C
Primary
1671
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DEFA6
Liquid
DEFA6
DEFA6 Fusion Protein Ag12341
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados