Learn More
Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators
Product Code. 13162053 Shop All Thermo Scientific Products
Change view
Click to view available options
Displacement:
116L/min. (at 60Hz); 97L/min. (at 50Hz)
195L/min. (at 60Hz); 162L/min. (at 50Hz)
283L/min. (at 60Hz); 237L/min. (at 50Hz)
76L/min. (at 60Hz); 62L/min. (at 50Hz)
Voltage:
230 V
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displacement Voltage Unit Size
13162053 116L/min. (at 60Hz); 97L/min. (at 50Hz) 230 V Each
10214832 195L/min. (at 60Hz); 162L/min. (at 50Hz) 230 V Each
10145182 283L/min. (at 60Hz); 237L/min. (at 50Hz) 230 V Each
10041442 76L/min. (at 60Hz); 62L/min. (at 50Hz) 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13162053 Supplier Thermo Scientific™ Supplier No. VLP120230
  • ONLINE EXCLUSIVE PRICE
    4285.00€
    3668.81€ / paquete de 1
    Ahorro 616.19€
    14% Descuento
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Achieve maximum solute recovery with the Thermo Scientific™Deep Vacuum Oil Pump.
Achieve maximum solute recovery with the Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators, designed to work with all Thermo Scientific™ SpeedVac™ concentrating and freeze drying systems.
Features:
  • Three-position gas ballast control optimizes pump performance to suit all applications
  • Large, sideview sight glass allows for convenient checking of proper oil levels at a glance
All Rotary Vane Oil Pumps come standard with:
  • Muffler (OME190 Oil Mist Eliminator)
  • Two Oil Filters (MF190)
  • Quick Drain Kit
TRUSTED_SUSTAINABILITY
Specifications
Description VLP120; 230V 50Hz
Type Deep Vacuum Oil Pump
Displacement 116L/min. (at 60Hz); 97L/min. (at 50Hz)
Oil Capacity 0.7 L
Ultimate Vacuum Level 1.5 mtorr/1.95 x10³ mbar
Vacuum Inlet Fitting 12.7 OD
Width (English) 6 in.
Width (Metric) 160 mm
Height (English) 9 in.
Height (Metric) 230 mm
Depth (English) 17 in.
Depth (Metric) 420 mm
Weight (English) 48 lb.
Weight (Metric) 22 kg
Electrical Requirements 230 V 50 Hz
Voltage 230 V
Frequency 50 Hz