missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DEDD2 Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP0311
Detalles adicionales : Peso : 0.01000kg
Descripción
DEDD2 Synthetic Peptide, PEP-0311, from Invitrogen.
This peptide corresponds to 11 amino acids near the amino terminus of human DEDD2.PEP-0311 can be used as a blocking peptide with polyclonal antibody PA5-20214.Especificaciones
162989 | |
purified | |
Human DEDD2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q8WXF8 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
DEDD2 | |
0.02% sodium azide | |
50 μg | |
PBS |