Learn More
Novus Biologicals™ DEDD Overexpression Lysate
Brand:  Novus Biologicals™ NBL1-09815
Additional Details : Peso : 0.00970kg
Product Code. 18215395
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock
Description
Description
The DEDD Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC259 and based on accession number NM_139711. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens death effector domain containing (DEDD), transcript variant 3, mRNA.
Native
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
For Research Use Only