missing translation for 'onlineSavingsMsg'
Learn More

Dectin-1/CLEC7A Antibody (010), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90491AF647

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213306

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

Dectin-1/CLEC7A
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified
Immunocytochemistry, Immunofluorescence
10
Immunocytochemistry/Immunofluorescence
Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Dectin-1/CLEC7A (NP_064392.2; Phe69-Leu244).
0.1 mL
Signal Transduction
64581
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales