Learn More
Decamethonium Iodide, Mikromol™
Product Code. 30050792
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
30050792 250 mg 250mg
1 options
This item is not returnable. View return policy
Product Code. 30050792 Supplier Mikromol Supplier No. MM3044.00
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Decamethonium Iodide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Decamethonium Iodide
CAS 1420-40-2
Analyte or Component Names Decamethonium Iodide
Shipping Condition Room Temperature
Molecular Formula C16 H38 N2 . 2 I
Certifications/Compliance ISO/IEC 17025
Synonym 1,10-Decanediaminium, N1,N1,N1,N10,N10,N10-hexamethyl-, iodide (1:2), 1,10-Decanediaminium, N,N,N,N',N',N'-hexamethyl-, diiodide (9CI), Ammonium, decamethylenebis[trimethyl- iodide] (6CI), Ammonium, decamethylenebis[trimethyl-, diiodide (8CI), Decamethylenebis(trimethylammonium) iodide (6CI), Decamethionium iodide, Decamethonium diiodide, Decamethonium iodide, Decamethylenebis(trimethylammonium) diiodide, Eulissin A, Eulixine, Procuran
InChI Formula InChI=1S/C16H38N2.2HI/c1-17(2,3)15-13-11-9-7-8-10-12-14-16-18(4,5)6;;/h7-16H2,1-6H3;2*1H/q+2;;/p-2
SMILES [I-].[I-].C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C
IUPAC Name trimethyl-[10-(trimethylazaniumyl)decyl]azanium;diiodide
Molecular Weight (g/mol) 512.3
Quantity 250 mg
Formula Weight 512.1124 g/mol