missing translation for 'onlineSavingsMsg'
Learn More

DDX6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 14632-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813374

  • 319.00€ / 150 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.
Especificaciones

Especificaciones

DDX6
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P26196, P54823
ATP dependent RNA helicase p54, DDX6, DEAD box protein 6, HLR2, Oncogene RCK, P54, RCK
Rabbit
IgG
150 μL
-20°C
Primary
13209, 1656, 500988
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
DDX6
Liquid
Ddx6
DDX6 Fusion Protein Ag6211
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados