missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DDX6 (Human) Recombinant Protein
Human DDX6 full-length ORF ( AAH65007, 1 a.a. - 483 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001656-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSTARTENPVIMGLSSQNGQLRGPVKPTGGPGGGGTQTQQQMNQLKNTNTINNGTQQQAQSMTTTIKPGDDWKKTLKLPPKDLRIKTSDVTSTKGNEFEDYCLKRELLMGIFEMGWEKPSPIQEESIPIALSGRDILARAKNGTGKSGAYLIPLLERLDLKKDNIQAMVIVPTRELALQVSQICIQVSKHMGGAKVMATTGGTNLRDDIMRLDDTVHVVIATPGRILDLIKKGVAKVDHVQMIVLDEADKLLSQDFVQIMEDIILTLPKNRQILLYSATFPLSVQKFMNSHLQKPYEINLMEELTLKGVTQYYAYVTERQKVHCLNTLFSRLQINQSIIFCNSSQRVELLAKKISQLGYSCFYIHAKMRQEHRNRVFHDFRNGLCRNLVCTDLFTRGIDIQAVNVVINFDFPKLAETYLHRIGRSGRFGHLGLAINLITYDDRFNLKSIEEQLGTEIKPIPSNIDKSLYVAEYHSEPVEDEKP
Especificaciones
AAH65007 | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDX6 | |
GST |
1656 | |
Wheat germ expression system | |
25 μg | |
FLJ36338, HLR2, P54, RCK | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |