missing translation for 'onlineSavingsMsg'
Learn More

DDX39B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 14798-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813624

  • 319.00€ / 150 microlitros

Envío estimado: 22-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody may cross-react with DDX39A.

This gene encodes a member of the DEAD box family of RNA-dependent ATPases that mediate ATP hydrolysis during pre-mRNA splicing. The encoded protein is an essential splicing factor required for association of U2 small nuclear ribonucleoprotein with pre-mRNA, and also plays an important role in mRNA export from the nucleus to the cytoplasm. A cluster of genes, BAT1-BAT5, is localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. Mutations in this gene may be associated with rheumatoid arthritis. Alternatively spliced transcript variants encoding the same protein have been described.
Especificaciones

Especificaciones

DDX39B
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13838, Q63413, Q9Z1N5
ATP dependent RNA helicase p47, BAT1, D6S81E, DDX39B, DEAD box protein UAP56, HLA B associated transcript 1, Spliceosome RNA helicase BAT1, UAP56
Rabbit
IgG
150 μL
-20°C
Primary
114612, 53817, 7919
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
DDX39B
Liquid
Ddx39b
DDX39B Fusion Protein Ag6512
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados