Learn More
DDX21 Mouse anti-Human, CoraLite 488, Clone: 1B9C8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66925-100UL
Detalles adicionales : Peso : 0.00010kg
Descripción
DDX21 protein belongs to DEAD box protein family which is characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD). As a putative RNA helicase, DDX21 unwinds double-stranded RNA, folds single-stranded RNA and is involved in process including ribosomal RNA biogeneis, RNA editing and general transcription. Interaction of DDX21 and c-Jun was reported in ribosomal RNA processing.
Especificaciones
DDX21 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig | |
IgG1 |
Immunofluorescence, Flow Cytometry | |
1B9C8 | |
CoraLite 488 | |
100 μL | |
9188 | |
Liquid |