missing translation for 'onlineSavingsMsg'
Learn More

DDRGK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-87256

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213919

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

DDRGK1 Polyclonal specifically detects DDRGK1 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

DDRGK1
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
65992
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
C20orf116, chromosome 20 open reading frame 116, DDRGK domain containing 1, DDRGK domain-containing protein 1, dJ1187M17.3, MGC2592
The immunogen is a synthetic peptide directed towards the N-terminal region of human DDRGK1
100 μL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales