missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR1 Human Protein
DDR1 Human Protein
Marca: Invitrogen PV6047
Detalles adicionales : Peso : 0.50000kg
Descripción
DDR1 is a receptor tyrosine kinase activated by collagen and is reported to regulate adhesion, cell migration and other cellular processes.Especificaciones
NP_001945 | |
Pharma and Biopharma, Protein Assays and Analysis, Kinase Assay | |
780kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
DDR1 | |
The ability of DDR1 to bind the ATP competitor Kinase Tracer 178 (PV5593) was demonstrated for this lot of DDR1 in the LanthaScreen™ Eu Kinase Binding Assay. Increasing amounts of Kinase Tracer 178 (PV5593) were incubated with 5 nM DDR1 and 2 nM Eu-anti-GST Antibody (PV5594) in Kinase Buffer A (PV3189) with and without the presence of 10μM dasatinib as the competitor for the Kinase Tracer | |
aa 440 to 876 | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
0.39mg/mL | |
780 | |
DDR1 Human Protein | |
–80°C | |
1 Each | |
DDR1 Human Protein | |
DDR kinase family | |
DDR1 Human Protein | |
GST Tag | |
Kinases and inhibitors | |
85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.