missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17758-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.Especificaciones
DCK | |
Polyclonal | |
Unconjugated | |
DCK | |
DCK, deoxycytidine kinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13178, 1633, 79127 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P27707, P43346, P48769 | |
Dck | |
DCK Fusion Protein Ag12123 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |