missing translation for 'onlineSavingsMsg'
Learn More

DC-LAMP Antibody (007), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89553DL594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214289

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

DC-LAMP
Monoclonal
DyLight 594
CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208
This antibody was obtained from a rabbit immunized with purified, recombinant Human DC-LAMP (NP_055213.2; Lys28-Thr381).
0.1 mL
Dendritic Cell Markers
27074
Store at 4°C in the dark.
IgG
Immunofluorescence
7
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales