missing translation for 'onlineSavingsMsg'
Learn More

DBT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 12451-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16820174

  • 319.00€ / 150 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The branched-chain alpha-keto acid dehydrogenase complex (BCKD) is an inner-mitochondrial enzyme complex involved in the breakdown of the branched-chain amino acids isoleucine, leucine, and valine. The BCKD complex is thought to be composed of a core of 24 transacylase (E2) subunits, and associated decarboxylase (E1), dehydrogenase (E3), and regulatory subunits. This gene encodes the transacylase (E2) subunit. Mutations in this gene result in maple syrup urine disease, type 2. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Especificaciones

Especificaciones

DBT
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P11182, P53395
DBT, E2, E2B
Rabbit
IgG
150 μL
-20°C
Primary
13171, 1629, 29611
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
DBT
Liquid
Dbt
DBT Fusion Protein Ag3101
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados