Learn More
Emma Safety Footwear Dave (Ruffneck) Safety Shoes
Looks and feels like a classic sneaker. Highly supportive reinforced ruff neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
Marca:  Emma Safety Footwear 914546-43
Detalles adicionales : Peso : 1.62000kg
 Ver más versiones de este producto
Código de producto. 15929071
  • 90.80€ / 1 par
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • S3-Safety shoe with sole plate for wet work
  • Nubuck / Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • ESD certified
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Especificaciones
Especificaciones
EN ISO 20345:2011, EN 12568:2010
Brown
Hydro-Tec™ Sanitized Silver
43
Laces
Dave (Ruffneck)
Nubuck, Oiled Nubuck
Steel Toe